Learn More
MSC Starrett Micrometer Case, Wood, For 300 mm Micrometer, 1 ea
Catalog No. 5000510250
Encompass
This case is designed for the protection and storage of 300 mm (12 in.) outside micrometers when they are not in use.
  • Constructed from wood
  • Compatible with series 216Z-12 and 436Z-12 outside micrometers
  • Designed for 300 mm (12 in.) micrometer sizes
  • Originates from the United States
Catalog No. 50-005-10250 Supplier MSC Supplier No. 86416328
This case is designed for the protection and storage of 300 mm (12 in.) outside micrometers when they are not in use.
  • Constructed from wood
  • Compatible with series 216Z-12 and 436Z-12 outside micrometers
  • Designed for 300 mm (12 in.) micrometer sizes
  • Originates from the United States