Learn More
MSC Starrett Combination Set, 24 in, Square, Center, Protractor Heads, 4R Graduations
Catalog No. 5000515955
Encompass
This combination set is a precision measuring tool designed for precise measurement and layout tasks. It includes a hardened steel blade and three interchangeable heads: a square head, a center head, and a reversible protractor head. The blade features a regular finish with 4R graduations, and the heads are constructed from durable cast iron.
  • Blade length: 24 inches
  • Includes square, center, and reversible protractor heads
  • Hardened steel blade and cast iron heads
  • Reversible lock bolts
  • Integrated scriber
  • Spirit levels in both the square head and the protractor head
  • Direct reading double 180-degree protractor scale
  • Manufactured in the USA
Catalog No. 50-005-15955 Supplier MSC Supplier No. 86414554
This combination set is a precision measuring tool designed for precise measurement and layout tasks. It includes a hardened steel blade and three interchangeable heads: a square head, a center head, and a reversible protractor head. The blade features a regular finish with 4R graduations, and the heads are constructed from durable cast iron.
  • Blade length: 24 inches
  • Includes square, center, and reversible protractor heads
  • Hardened steel blade and cast iron heads
  • Reversible lock bolts
  • Integrated scriber
  • Spirit levels in both the square head and the protractor head
  • Direct reading double 180-degree protractor scale
  • Manufactured in the USA