Learn More
MSC Starrett Combination Square, 12 in Blade, Square Head, 16R
Catalog No. 5000528966
Encompass
This combination square with square head includes a reversible lock bolt, scriber, spirit level, and a hardened steel, photo-engraved blade with satin chrome finish. The square heads are forged hardened steel with smooth, black enamel finish. It features a 12 in blade with 16R graduations.
  • Reversible lock bolt included
  • Scriber included
  • Spirit level included
  • Hardened steel, machine divided blade
  • Satin chrome blade finish
  • Forged and hardened steel head with smooth black enamel finish
  • Blade length: 12 in
  • Graduation type: 16R
  • Graduations: quick reading 32nds, 64ths, aircraft quick reading 50ths, 100ths
  • Head type: square
  • Made in the United States
  • Number of pieces: 2
Catalog No. 50-005-28966 Supplier MSC Supplier No. 86402211
This combination square with square head includes a reversible lock bolt, scriber, spirit level, and a hardened steel, photo-engraved blade with satin chrome finish. The square heads are forged hardened steel with smooth, black enamel finish. It features a 12 in blade with 16R graduations.
  • Reversible lock bolt included
  • Scriber included
  • Spirit level included
  • Hardened steel, machine divided blade
  • Satin chrome blade finish
  • Forged and hardened steel head with smooth black enamel finish
  • Blade length: 12 in
  • Graduation type: 16R
  • Graduations: quick reading 32nds, 64ths, aircraft quick reading 50ths, 100ths
  • Head type: square
  • Made in the United States
  • Number of pieces: 2