Learn More
MSC Starrett Combination Square, 12 in, Square Head, 16R Graduation, Cast Iron Head
Catalog No. 5000537320
Encompass
This combination square is designed for precise measurements and marking. It features a hardened steel blade with a regular finish and a cast iron head, making it suitable for various workshop and professional applications.
  • Reversible lock bolt
  • Built-in scriber
  • Integrated spirit level
  • Hardened steel, machine divided blade
  • Cast iron head with black wrinkle finish
  • 12 in blade length
  • 16R graduation type
  • Graduations include quick reading 32nds, 64ths, aircraft quick reading 50ths, and 100ths
Catalog No. 50-005-37320 Supplier MSC Supplier No. 86400470
This combination square is designed for precise measurements and marking. It features a hardened steel blade with a regular finish and a cast iron head, making it suitable for various workshop and professional applications.
  • Reversible lock bolt
  • Built-in scriber
  • Integrated spirit level
  • Hardened steel, machine divided blade
  • Cast iron head with black wrinkle finish
  • 12 in blade length
  • 16R graduation type
  • Graduations include quick reading 32nds, 64ths, aircraft quick reading 50ths, and 100ths