Learn More
MSC Starrett Combination Square Blade, 4 Inch, 4R Graduation, Satin Chrome Finish
Catalog No. 5000514076
Encompass
This combination square blade is a high-quality replacement or accessory blade designed for use with Junior Combination Squares. Crafted from hardened steel, this 4-inch blade features precise 4R graduations, making it an essential tool for mechanics, toolmakers, and patternmakers who require accurate measurements in 8ths, 16ths, quick-reading 32nds, and 64ths of an " Its satin chrome finish provides durability and readability.
  • Designed for use with Junior Combination Squares.
  • Constructed from hardened steel for durability.
  • Features 4R graduations for precise measurements.
  • Measurements include 8ths, 16ths, quick-reading 32nds, and 64ths.
  • Satin chrome finish enhances readability and corrosion resistance.
  • Ideal for mechanics, toolmakers, and patternmakers.
Catalog No. 50-005-14076 Supplier MSC Supplier No. 58694175
This combination square blade is a high-quality replacement or accessory blade designed for use with Junior Combination Squares. Crafted from hardened steel, this 4-inch blade features precise 4R graduations, making it an essential tool for mechanics, toolmakers, and patternmakers who require accurate measurements in 8ths, 16ths, quick-reading 32nds, and 64ths of an " Its satin chrome finish provides durability and readability.
  • Designed for use with Junior Combination Squares.
  • Constructed from hardened steel for durability.
  • Features 4R graduations for precise measurements.
  • Measurements include 8ths, 16ths, quick-reading 32nds, and 64ths.
  • Satin chrome finish enhances readability and corrosion resistance.
  • Ideal for mechanics, toolmakers, and patternmakers.