Learn More
MSC Starrett Combination Square Set, 2 Piece, 12 in, 16R
Catalog No. 5000527628
Encompass
This combination square set features a 2-piece design and a 12-inch blade, ideal for precision measurement and layout tasks. It includes a durable steel blade with 16R graduation and a robust cast iron head, making it suitable for a variety of workshop and inspection applications.
  • 2-piece design for versatility
  • 12 in steel blade for accurate measurements
  • 16R graduation for detailed marking
  • Cast iron head for durability and stability
  • Suitable for measurement and layout tasks
Catalog No. 50-005-27628 Supplier MSC Supplier No. 86400652
This combination square set features a 2-piece design and a 12-inch blade, ideal for precision measurement and layout tasks. It includes a durable steel blade with 16R graduation and a robust cast iron head, making it suitable for a variety of workshop and inspection applications.
  • 2-piece design for versatility
  • 12 in steel blade for accurate measurements
  • 16R graduation for detailed marking
  • Cast iron head for durability and stability
  • Suitable for measurement and layout tasks