Learn More
MSC Starrett Depth Gage Attachment, 6 in Rod Length, for Height Gages
Catalog No. 5000513148
Encompass
This depth gage attachment is designed to enhance the capabilities of Starrett height gages by replacing standard scribers. It allows for precise measurement of depths in holes, slots, and recesses, as well as over high projections. Featuring adjustable rods secured by a knurled binding nut, its 6 in (150 mm) rod length and radiused ends ensure accurate point contact for diverse applications.
  • Replaces standard scribers for height gages.
  • Measures depths of holes, slots, and recesses.
  • Capable of measuring over high projections.
  • Equipped with adjustable rods.
  • Rods secured by a knurled binding nut.
  • Rod ends have a slight radius for precise point contact.
  • Provides 6 in (150 mm) rod length.
  • Designed for use with height gages.
Catalog No. 50-005-13148 Supplier MSC Supplier No. 09770959
This depth gage attachment is designed to enhance the capabilities of Starrett height gages by replacing standard scribers. It allows for precise measurement of depths in holes, slots, and recesses, as well as over high projections. Featuring adjustable rods secured by a knurled binding nut, its 6 in (150 mm) rod length and radiused ends ensure accurate point contact for diverse applications.
  • Replaces standard scribers for height gages.
  • Measures depths of holes, slots, and recesses.
  • Capable of measuring over high projections.
  • Equipped with adjustable rods.
  • Rods secured by a knurled binding nut.
  • Rod ends have a slight radius for precise point contact.
  • Provides 6 in (150 mm) rod length.
  • Designed for use with height gages.