Learn More
MSC Starrett Depth Micrometer, 0-3 inch, 3 Rods, 4 inch Base, 0.001 inch Graduation
Catalog No. 5000514210
Encompass
This mechanical depth micrometer is designed for precise measurement of depths of holes, slots, shoulders, and projections. It features a 0-3 inch measuring range with a 0.001 inch graduation for accurate readings. The tool is equipped with a 4 inch base and three 5/32 inch diameter measuring rods, providing versatility for various applications. Its satin chrome finish offers glare-free reading and resistance to rust, ensuring durability and consistent performance.
  • Combination ratchet and speeder for uniform pressure and quick adjustment.
  • Ring-type knurled lock nut for quick and secure locking.
  • Satin chrome reading surface is glare-free and resists rust.
  • Clear, easily read white numbers on a black background to reduce errors.
  • Includes a padded case for protection and storage.
Catalog No. 50-005-14210 Supplier MSC Supplier No. 86417649
This mechanical depth micrometer is designed for precise measurement of depths of holes, slots, shoulders, and projections. It features a 0-3 inch measuring range with a 0.001 inch graduation for accurate readings. The tool is equipped with a 4 inch base and three 5/32 inch diameter measuring rods, providing versatility for various applications. Its satin chrome finish offers glare-free reading and resistance to rust, ensuring durability and consistent performance.
  • Combination ratchet and speeder for uniform pressure and quick adjustment.
  • Ring-type knurled lock nut for quick and secure locking.
  • Satin chrome reading surface is glare-free and resists rust.
  • Clear, easily read white numbers on a black background to reduce errors.
  • Includes a padded case for protection and storage.