Learn More
MSC Starrett Depth Micrometer Replacement Rod, 0-1 Inch, For 449 Series
Catalog No. 5000531337
Encompass
This replacement rod for depth micrometers is engineered for precision and durability. It offers a measurement range of 0 to 1 inch (0 to 25 mm) and is specifically designed for compatibility with 449 Series micrometer depth gages. Manufactured with high standards, it ensures consistent and accurate performance.
  • Adjustable rods are hardened, ground, and lapped for permanent accuracy.
  • Manufactured in the USA with US and global content.
  • Designed for use with 449 Series micrometer depth gages.
  • Provides a measurement range of 0 to 1 inch (0 to 25 mm).
Catalog No. 50-005-31337 Supplier MSC Supplier No. 86418431
This replacement rod for depth micrometers is engineered for precision and durability. It offers a measurement range of 0 to 1 inch (0 to 25 mm) and is specifically designed for compatibility with 449 Series micrometer depth gages. Manufactured with high standards, it ensures consistent and accurate performance.
  • Adjustable rods are hardened, ground, and lapped for permanent accuracy.
  • Manufactured in the USA with US and global content.
  • Designed for use with 449 Series micrometer depth gages.
  • Provides a measurement range of 0 to 1 inch (0 to 25 mm).