Learn More
MSC Starrett Dial Caliper, 6 inch, 0.001 inch Graduation, Black Dial
Catalog No. 5000510253
Encompass
This American-made dial caliper is widely used by mechanics and toolmakers, offering direct reading, ease of handling, reliability, and accuracy. It features a black dial with a 0-6 inch range and 0.001 inch graduations, constructed from hardened stainless steel for durability.
  • American made
  • Direct reading, reliable, and accurate measurements
  • Sharp, clear dial graduations
  • Sharp, black graduations on satin finished bar
  • Knife-edge contacts for inside and outside measurements
  • One-hand use with thumb-operated fine adjustment roll
  • Lock screw for dial bezel and for holding sliding jaw in position
  • Hardened stainless steel bar, measuring surfaces, rack, gears, and depth rod
  • Positive, split-gear anti-backlash control
  • Covered rack depth rod
Catalog No. 50-005-10253 Supplier MSC Supplier No. 86435047
This American-made dial caliper is widely used by mechanics and toolmakers, offering direct reading, ease of handling, reliability, and accuracy. It features a black dial with a 0-6 inch range and 0.001 inch graduations, constructed from hardened stainless steel for durability.
  • American made
  • Direct reading, reliable, and accurate measurements
  • Sharp, clear dial graduations
  • Sharp, black graduations on satin finished bar
  • Knife-edge contacts for inside and outside measurements
  • One-hand use with thumb-operated fine adjustment roll
  • Lock screw for dial bezel and for holding sliding jaw in position
  • Hardened stainless steel bar, measuring surfaces, rack, gears, and depth rod
  • Positive, split-gear anti-backlash control
  • Covered rack depth rod