Learn More
MSC Starrett Dial Drop Indicator, 4.0000 in, 0-100 Dial, Lug Back
Catalog No. 5000531583
Encompass
This dial drop indicator is a precision measuring tool designed for accurate readings in various applications. It features a robust construction with specific design elements for reliable performance and long-lasting use.
  • Max measurement of 4.0000 in
  • Dial graduation of 0.001000 in
  • 0-100 dial reading
  • Lug back
  • Jeweled bearing
  • White dial color
  • Revolution counter
  • No-glare satin finish
  • Stainless steel stem
  • Unbreakable crystal
  • Rugged design
  • Includes flat back
Catalog No. 50-005-31583 Supplier MSC Supplier No. 86427366
This dial drop indicator is a precision measuring tool designed for accurate readings in various applications. It features a robust construction with specific design elements for reliable performance and long-lasting use.
  • Max measurement of 4.0000 in
  • Dial graduation of 0.001000 in
  • 0-100 dial reading
  • Lug back
  • Jeweled bearing
  • White dial color
  • Revolution counter
  • No-glare satin finish
  • Stainless steel stem
  • Unbreakable crystal
  • Rugged design
  • Includes flat back