Learn More
MSC Starrett Dial Indicator, 0.025 in Range, 0.0001 in Graduations, Jewel Bearing
Catalog No. 5000531581
Encompass
This dial indicator offers precise measurements with its rugged and simple unit construction. It features a hardened steel gear train and jewel bearings for sensitive and durable operation, and comes with both a lug-on-center back and a flat back for versatile mounting.
  • Sharp bezel serrations for positive grip.
  • Unbreakable crystal.
  • Hardened stainless steel stem.
  • No-glare satin finish on case.
  • Hardened stainless steel rack and spindle.
  • Massive bridge for rigid bearing support.
  • Hardened stainless steel gears and pinions.
  • Non-reflecting white eggshell finish on dial.
  • Jewel bearings.
  • Lug on center back installed, flat back included.
  • Optional antimagnetic and special non-shock mechanisms.
Catalog No. 50-005-31581 Supplier MSC Supplier No. 86426897
This dial indicator offers precise measurements with its rugged and simple unit construction. It features a hardened steel gear train and jewel bearings for sensitive and durable operation, and comes with both a lug-on-center back and a flat back for versatile mounting.
  • Sharp bezel serrations for positive grip.
  • Unbreakable crystal.
  • Hardened stainless steel stem.
  • No-glare satin finish on case.
  • Hardened stainless steel rack and spindle.
  • Massive bridge for rigid bearing support.
  • Hardened stainless steel gears and pinions.
  • Non-reflecting white eggshell finish on dial.
  • Jewel bearings.
  • Lug on center back installed, flat back included.
  • Optional antimagnetic and special non-shock mechanisms.