Learn More
MSC Starrett Dial Indicator, 1.000 inch Range, 0.001 inch Graduations, 0-100 Dial, Jewel Bearing
Catalog No. 5000536658
Encompass
This dial indicator features a shockless, hardened steel gear train and jewel bearings, supplied with a lug-on-center back. Antimagnetic mechanisms are optional, and a special non-shock mechanism is available. This model offers a 1.000 inch range with 0-100 dial reading and 0.001 inch graduations, including a case stem cap for lever control.
  • Rugged unit construction and universal design
  • Sharp bezel serrations for positive grip
  • Unbreakable crystal
  • Hardened stainless steel stem, rack, and spindle
  • No-glare satin finish on case
  • Massive bridge for rigid bearing support
  • Hardened stainless steel gears and pinions
  • Non-reflecting white eggshell finish on dial
  • Jewel bearings
  • Case stem cap supplied as standard
  • Lug on center back installed, with flat back included
Catalog No. 50-005-36658 Supplier MSC Supplier No. 90607045
This dial indicator features a shockless, hardened steel gear train and jewel bearings, supplied with a lug-on-center back. Antimagnetic mechanisms are optional, and a special non-shock mechanism is available. This model offers a 1.000 inch range with 0-100 dial reading and 0.001 inch graduations, including a case stem cap for lever control.
  • Rugged unit construction and universal design
  • Sharp bezel serrations for positive grip
  • Unbreakable crystal
  • Hardened stainless steel stem, rack, and spindle
  • No-glare satin finish on case
  • Massive bridge for rigid bearing support
  • Hardened stainless steel gears and pinions
  • Non-reflecting white eggshell finish on dial
  • Jewel bearings
  • Case stem cap supplied as standard
  • Lug on center back installed, with flat back included