Learn More
MSC Starrett Dial Test Indicator, 0.001 inch Graduation, 0.060 inch Range, Black Dial, Swivel Head
Catalog No. 5000531599
Encompass
The Starrett 811 Dial Test Indicator with Swivel Head provides precise measurements for laboratory and industrial applications. Its swivel head allows positioning from horizontal to vertical, or any angle up to 90 degrees. Featuring a smooth, jeweled movement and a large 1-3/8 inch (35 mm) dial diameter, this indicator ensures readability and reliable performance.
  • Contact point reverses automatically, maintaining clockwise hand rotation.
  • Contacts are frictionally adjustable and replaceable.
  • Swivel head allows versatile positioning for line of sight.
  • Smooth, jeweled movement for consistent operation.
  • Large 1-3/8 inch (35 mm) dial diameter enhances readability.
  • Includes various attachments for versatile mounting.
  • Two positioning mounts compatible with dovetail test indicator accessories.
Catalog No. 50-005-31599 Supplier MSC Supplier No. 86435807
The Starrett 811 Dial Test Indicator with Swivel Head provides precise measurements for laboratory and industrial applications. Its swivel head allows positioning from horizontal to vertical, or any angle up to 90 degrees. Featuring a smooth, jeweled movement and a large 1-3/8 inch (35 mm) dial diameter, this indicator ensures readability and reliable performance.
  • Contact point reverses automatically, maintaining clockwise hand rotation.
  • Contacts are frictionally adjustable and replaceable.
  • Swivel head allows versatile positioning for line of sight.
  • Smooth, jeweled movement for consistent operation.
  • Large 1-3/8 inch (35 mm) dial diameter enhances readability.
  • Includes various attachments for versatile mounting.
  • Two positioning mounts compatible with dovetail test indicator accessories.