Learn More
MSC Starrett Dial Test Indicator, Swivel Head, 0.060 in Range, 0.001 in Graduations, White Dial
Catalog No. 5000530246
Encompass
The Starrett 811 Dial Test Indicator with Swivel Head is a versatile instrument designed for precise measurements. It features a unique swivel head that allows for flexible positioning from horizontal to vertical and any angle up to 90 degrees. With a 0.060 in range and 0.001 in graduations, it ensures accurate readings on its 0-30-0 white dial. This indicator comes as a complete kit with several useful attachments.
  • Swivel head allows versatile positioning from horizontal to vertical and any angle in between.
  • Contact point reverses automatically, always maintaining clockwise hand rotation.
  • Contacts are frictionally adjustable and replaceable.
  • Features smooth, jeweled movement.
  • Large 1-3/8 in dial diameter for increased readability.
  • Includes a dovetail body clamp, tool post holder, swivel post snug with dovetail indicator clamp, and a snug and rod unit.
Catalog No. 50-005-30246 Supplier MSC Supplier No. 86436425
The Starrett 811 Dial Test Indicator with Swivel Head is a versatile instrument designed for precise measurements. It features a unique swivel head that allows for flexible positioning from horizontal to vertical and any angle up to 90 degrees. With a 0.060 in range and 0.001 in graduations, it ensures accurate readings on its 0-30-0 white dial. This indicator comes as a complete kit with several useful attachments.
  • Swivel head allows versatile positioning from horizontal to vertical and any angle in between.
  • Contact point reverses automatically, always maintaining clockwise hand rotation.
  • Contacts are frictionally adjustable and replaceable.
  • Features smooth, jeweled movement.
  • Large 1-3/8 in dial diameter for increased readability.
  • Includes a dovetail body clamp, tool post holder, swivel post snug with dovetail indicator clamp, and a snug and rod unit.