Learn More
MSC Starrett Dial Thickness Gage, 0 to 3/8 in. Measurement, 0.001 in. Graduation, 1/2 in. Throat Depth
Catalog No. 5000534274
Encompass
This Starrett Dial Thickness Gage offers precise measurement capabilities for various applications.
  • Handy pocket gage
  • Chrome plated case
  • Unbreakable crystal dial cover
  • Small count hand
  • Traceability certification included: California PROP 65
Catalog No. 50-005-34274 Supplier MSC Supplier No. 00126144
This Starrett Dial Thickness Gage offers precise measurement capabilities for various applications.
  • Handy pocket gage
  • Chrome plated case
  • Unbreakable crystal dial cover
  • Small count hand
  • Traceability certification included: California PROP 65