Learn More
MSC Starrett DIGI-CHEK Height Gage, 0.1 to 12.1 in, 0.0002 in Accuracy
Catalog No. 5000511803
Encompass
The Starrett DIGI-CHEK Height Gage provides precise measurements, combining the accuracy of gage blocks with a digital readout and micrometer head for versatile applications.
  • Combines gage block accuracy with a precision micrometer head and digital readout.
  • Digital readout offers 0.001 in (0.01 mm) resolution and a 1 in (25 mm) range.
  • Reads directly from the micrometer head to 0.0001 in (0.002 mm).
  • Micrometer head with a speeder knob for rapid positioning.
  • Enables checking of both over and under heights in a single setting.
  • Features a heavily flanged frame and tip-proof base for stability.
  • Free-standing gage block system conforms to temperature variations.
  • Highly accurate micrometer spindle with hardened, stabilized measuring threads.
Catalog No. 50-005-11803 Supplier MSC Supplier No. 86411873
The Starrett DIGI-CHEK Height Gage provides precise measurements, combining the accuracy of gage blocks with a digital readout and micrometer head for versatile applications.
  • Combines gage block accuracy with a precision micrometer head and digital readout.
  • Digital readout offers 0.001 in (0.01 mm) resolution and a 1 in (25 mm) range.
  • Reads directly from the micrometer head to 0.0001 in (0.002 mm).
  • Micrometer head with a speeder knob for rapid positioning.
  • Enables checking of both over and under heights in a single setting.
  • Features a heavily flanged frame and tip-proof base for stability.
  • Free-standing gage block system conforms to temperature variations.
  • Highly accurate micrometer spindle with hardened, stabilized measuring threads.