This electronic caliper offers precise measurements in industrial and laboratory settings. Made from hardened stainless steel, it provides durability and IP67 protection against dust and water. Featuring a large, easy-to-read LCD, it delivers accurate measurements with .0005 in (0.01 mm) resolution over a 0-6 in (150 mm) range.
- IP67 protection from coolants, water, chips, dust, and dirt.
- Hardened stainless steel construction.
- Large, high-contrast LCD readout.
- Resolution: .0005 in (0.01 mm).
- Measurement range: 0-6 in (150 mm).
- SPC output for data collection.
- Inch/millimeter conversion.
- Zero at any position; auto-off after 30 minutes.
- Display reactivates with movement, no loss of position.
- Includes redemption for a standard letter of certification.
- Comes with a protective case.
- Powered by an included CR2032 lithium battery.