Learn More
MSC Starrett Electronic Caliper, 0-6 inch, 0.0005 inch Resolution, Stainless Steel
Catalog No. 5000528400
Encompass
This electronic caliper is designed for precise measurements in various applications. Constructed from durable stainless steel, it offers a measuring range of 0-6 inches with a high resolution of 0.0005 inch, suitable for inside diameter, outside diameter, depth, and step measurements.
  • Constructed from hardened stainless steel for durability.
  • Provides precise measurements for inside diameter, outside diameter, depth, and step applications.
  • Features a knife-edge jaw type for accurate readings.
  • Powered by a CR2032 lithium battery, included for immediate use.
  • NIST traceability certified, ensuring measurement accuracy.
  • Includes a depth measurement rod for versatile use.
  • Comes with a protective case for storage and transport.
Catalog No. 50-005-28400 Supplier MSC Supplier No. 41709445
This electronic caliper is designed for precise measurements in various applications. Constructed from durable stainless steel, it offers a measuring range of 0-6 inches with a high resolution of 0.0005 inch, suitable for inside diameter, outside diameter, depth, and step measurements.
  • Constructed from hardened stainless steel for durability.
  • Provides precise measurements for inside diameter, outside diameter, depth, and step applications.
  • Features a knife-edge jaw type for accurate readings.
  • Powered by a CR2032 lithium battery, included for immediate use.
  • NIST traceability certified, ensuring measurement accuracy.
  • Includes a depth measurement rod for versatile use.
  • Comes with a protective case for storage and transport.