Learn More
MSC Starrett End Measuring Rod, 17 inch, Insulated Handle, Spherical Ends
Catalog No. 5000531269
Encompass
This end measuring rod is designed for precise measurement tasks, ideal for checking and setting micrometers of 2 inch capacity and larger, comparing gages, and measuring parallel surfaces. It is constructed from special tool steel with hardened, accurately lapped spherical ends to ensure accuracy and durability.
  • Features an insulated handle to minimize expansion caused by body heat during use.
  • Ends are hardened and accurately lapped to a spherical radius for precise measurements.
  • Constructed from special tool steel for durability and stability.
  • Suitable for checking and setting micrometers of 2 inch capacity and larger.
  • Can be used on machine tools for comparing gages and checking precision measuring tools.
  • Useful for measuring parallel surfaces in various applications.
Catalog No. 50-005-31269 Supplier MSC Supplier No. 81532491
This end measuring rod is designed for precise measurement tasks, ideal for checking and setting micrometers of 2 inch capacity and larger, comparing gages, and measuring parallel surfaces. It is constructed from special tool steel with hardened, accurately lapped spherical ends to ensure accuracy and durability.
  • Features an insulated handle to minimize expansion caused by body heat during use.
  • Ends are hardened and accurately lapped to a spherical radius for precise measurements.
  • Constructed from special tool steel for durability and stability.
  • Suitable for checking and setting micrometers of 2 inch capacity and larger.
  • Can be used on machine tools for comparing gages and checking precision measuring tools.
  • Useful for measuring parallel surfaces in various applications.