Learn More
MSC Starrett Feeler Gage, 0.002 in Thick, 12 in Long, Parallel, Stainless Steel
Catalog No. 5000527123
Encompass
This feeler gage is designed for precise measurements and features a thickness of 0.002 " It is 12 inches long and made from durable stainless steel, ensuring reliability and accuracy in various applications. The parallel leaf design provides consistent and accurate gap measurement.
  • 0.002 in thickness
  • Parallel leaf style for consistent measurements
  • 12 in length for extended reach
  • 0.5 in width
  • Constructed from durable stainless steel
  • Made in China
Catalog No. 50-005-27123 Supplier MSC Supplier No. 39864996
This feeler gage is designed for precise measurements and features a thickness of 0.002 " It is 12 inches long and made from durable stainless steel, ensuring reliability and accuracy in various applications. The parallel leaf design provides consistent and accurate gap measurement.
  • 0.002 in thickness
  • Parallel leaf style for consistent measurements
  • 12 in length for extended reach
  • 0.5 in width
  • Constructed from durable stainless steel
  • Made in China