Learn More
MSC Starrett Feeler Gage Set, 108 Piece, Tempered Steel, 0.0015-0.015 in, Parallel
Catalog No. 5000516771
Encompass
The Starrett feeler gage set is a 108-piece assortment designed for precise measurement of small gaps and clearances. Made from tempered steel, these parallel feeler gages offer a thickness range from 0.0015 to 0.015 inches, making them suitable for a variety of industrial and scientific applications requiring accurate spacing.
  • Contains 108 individual leaves for a comprehensive range of measurements.
  • Features durable tempered steel construction for longevity and accuracy.
  • Parallel leaf style ensures consistent measurements across the length of the gage.
  • Leaves are 1/2 in wide by 12 in long, suitable for various applications.
  • Each gage is individually enveloped, and sets are boxed for organized storage.
Catalog No. 50-005-16771 Supplier MSC Supplier No. 06198014
The Starrett feeler gage set is a 108-piece assortment designed for precise measurement of small gaps and clearances. Made from tempered steel, these parallel feeler gages offer a thickness range from 0.0015 to 0.015 inches, making them suitable for a variety of industrial and scientific applications requiring accurate spacing.
  • Contains 108 individual leaves for a comprehensive range of measurements.
  • Features durable tempered steel construction for longevity and accuracy.
  • Parallel leaf style ensures consistent measurements across the length of the gage.
  • Leaves are 1/2 in wide by 12 in long, suitable for various applications.
  • Each gage is individually enveloped, and sets are boxed for organized storage.