Learn More
MSC Starrett Feeler Gage Set, 31 Leaves, Stainless Steel, 0.0015-0.0350 in Range
Catalog No. 5000537585
Encompass
This feeler gage set features 31 stainless steel leaves, offering a precise thickness range of 0.0015 to 0.0350 inches. Designed for various measurement applications, its durable construction ensures longevity and resistance to corrosion.
  • Composed of 31 leaves for a wide range of measurements.
  • Constructed from durable stainless steel for longevity and corrosion resistance.
  • Features a parallel leaf style for accurate gap measurement.
  • Each leaf has a length of 3-1/32 in and a width of 1/2 in.
  • Includes thicknesses ranging from 0.0015 to 0.0350 in.
  • Has both standard and metric markings.
  • NIST certification is available upon request.
Catalog No. 50-005-37585 Supplier MSC Supplier No. 31209257
This feeler gage set features 31 stainless steel leaves, offering a precise thickness range of 0.0015 to 0.0350 inches. Designed for various measurement applications, its durable construction ensures longevity and resistance to corrosion.
  • Composed of 31 leaves for a wide range of measurements.
  • Constructed from durable stainless steel for longevity and corrosion resistance.
  • Features a parallel leaf style for accurate gap measurement.
  • Each leaf has a length of 3-1/32 in and a width of 1/2 in.
  • Includes thicknesses ranging from 0.0015 to 0.0350 in.
  • Has both standard and metric markings.
  • NIST certification is available upon request.