Learn More
MSC Starrett Feeler Gage, Stainless Steel, 0.030 in Thick, 12 in Length
Catalog No. 5000533605
Encompass
These gages are widely used in automotive, aviation, diesel, and farm equipment manufacture and service, as well as in jig, fixture, gage, and experimental work. This specific product is 0.030 in thick and 12 in length.
  • Comes in an individual envelope.
  • Features a convenient 3/16 in (5 mm) hole punched in the end for hanging.
  • Rounded ends ensure easier handling.
  • Marked every 6 in (150 mm) with thickness for easy identification.
  • Made of high-quality stainless steel.
Catalog No. 50-005-33605 Supplier MSC Supplier No. 39023296
These gages are widely used in automotive, aviation, diesel, and farm equipment manufacture and service, as well as in jig, fixture, gage, and experimental work. This specific product is 0.030 in thick and 12 in length.
  • Comes in an individual envelope.
  • Features a convenient 3/16 in (5 mm) hole punched in the end for hanging.
  • Rounded ends ensure easier handling.
  • Marked every 6 in (150 mm) with thickness for easy identification.
  • Made of high-quality stainless steel.