Learn More
MSC Starrett Feeler Stock, Stainless Steel, 0.002 inch Thick, 1/2 inch Wide
Catalog No. 5000528413
Encompass
This stainless thickness gage, also known as feeler stock, is a precision tool used for measuring and setting small clearances. It is made from high-quality stainless steel and comes in a convenient rewindable dispenser case. It is ideal for adjusting tappets, spark plugs, distributor points, checking bearing clearances and gear play, fitting pistons, rings and pins, gaging narrow slots, and for shimming in fixturing and die work.
  • Made from high-quality stainless steel for durability and corrosion resistance.
  • Provided in a compact, sturdy plastic rewindable dispenser case for easy storage and use.
  • Each roll is marked every 6 in (150 mm) with a line for accurate and waste-free dispensing.
  • Offers a thickness of 0.002 in, a width of 1/2 in, and a total length of 25 ft.
  • Allows users to snip off the desired length without any waste.
Catalog No. 50-005-28413 Supplier MSC Supplier No. 39865092
This stainless thickness gage, also known as feeler stock, is a precision tool used for measuring and setting small clearances. It is made from high-quality stainless steel and comes in a convenient rewindable dispenser case. It is ideal for adjusting tappets, spark plugs, distributor points, checking bearing clearances and gear play, fitting pistons, rings and pins, gaging narrow slots, and for shimming in fixturing and die work.
  • Made from high-quality stainless steel for durability and corrosion resistance.
  • Provided in a compact, sturdy plastic rewindable dispenser case for easy storage and use.
  • Each roll is marked every 6 in (150 mm) with a line for accurate and waste-free dispensing.
  • Offers a thickness of 0.002 in, a width of 1/2 in, and a total length of 25 ft.
  • Allows users to snip off the desired length without any waste.