Learn More
MSC Starrett Gage Block, Rectangular, Steel, .149 in, Grade A1
Catalog No. 5000532305
Encompass
This individual rectangular gage block is designed for precise measurement and inspection applications. It features a nominal size of .149 inches and an accuracy grade of A1, ensuring high precision for critical measurements.
  • Rectangular steel construction
  • Nominal size of .149 in
  • Accuracy grade of A1
  • B89.1.9 accuracy grade 0
  • Suitable for inspection applications
Catalog No. 50-005-32305 Supplier MSC Supplier No. 40987463
This individual rectangular gage block is designed for precise measurement and inspection applications. It features a nominal size of .149 inches and an accuracy grade of A1, ensuring high precision for critical measurements.
  • Rectangular steel construction
  • Nominal size of .149 in
  • Accuracy grade of A1
  • B89.1.9 accuracy grade 0
  • Suitable for inspection applications