Learn More
MSC Starrett Gage Block Set, Steel, 81 pc, Grade 0, Square
Catalog No. 5000513628
Encompass
This steel gage block set is a precision measuring tool designed for inspection applications. It includes 81 blocks and is furnished in a protective wooden case.
  • Made from high-grade steel
  • Square shape
  • 81 blocks per set
  • Accuracy grade 0 (ASME B89.1.9-2002)
  • Measuring range of 0.05 to 4 in
  • Includes NIST & NVLAP traceability certification
  • Furnished in a wooden case
Catalog No. 50-005-13628 Supplier MSC Supplier No. 00348169
This steel gage block set is a precision measuring tool designed for inspection applications. It includes 81 blocks and is furnished in a protective wooden case.
  • Made from high-grade steel
  • Square shape
  • 81 blocks per set
  • Accuracy grade 0 (ASME B89.1.9-2002)
  • Measuring range of 0.05 to 4 in
  • Includes NIST & NVLAP traceability certification
  • Furnished in a wooden case