Learn More
MSC Starrett Gage Block Set, Steel, Grade 0, 81 Pc.
Catalog No. 5000516838
Encompass
This rectangular inch gage block set provides high-precision measurement capability with a range of 0.05 in. to 4 in. The 81-piece set is constructed from high-grade steel, offering the durability required for standard shop floor environments. Each set complies with ASME B89.1.9-2002 standards and is supplied with NIST and NVLAP traceability certification to ensure measurement accuracy.
  • Includes 81 individual blocks
  • Measurement range of 0.05 in. to 4 in.
  • Constructed from traditional high-grade steel
  • Accuracy grade 0
  • Supplied in a rugged wood storage case
  • Certified with NIST and NVLAP traceability
  • Rectangular block shape
Catalog No. 50-005-16838 Supplier MSC Supplier No. 06460877
This rectangular inch gage block set provides high-precision measurement capability with a range of 0.05 in. to 4 in. The 81-piece set is constructed from high-grade steel, offering the durability required for standard shop floor environments. Each set complies with ASME B89.1.9-2002 standards and is supplied with NIST and NVLAP traceability certification to ensure measurement accuracy.
  • Includes 81 individual blocks
  • Measurement range of 0.05 in. to 4 in.
  • Constructed from traditional high-grade steel
  • Accuracy grade 0
  • Supplied in a rugged wood storage case
  • Certified with NIST and NVLAP traceability
  • Rectangular block shape