Learn More
MSC Starrett Horizontal Dial Test Indicator, 0.80 mm Max, 0-40-0 Dial, Jeweled Bearing
Catalog No. 5000513412
Encompass
The Starrett Series 3909 Horizontal Dial Test Indicator is designed for precision measurements, featuring a robust gear-driven design and jeweled movement for accuracy. Its durable satin chrome finish ensures longevity, and the adjustable, replaceable contact point enhances versatility. With a high-contrast dial and automatic contact point reversal, this indicator provides reliable performance for various laboratory and industrial applications.
  • Precision gear-driven design
  • Jeweled movement
  • Satin chrome finish
  • Adjustable contact
  • Durable construction
  • Replaceable contact
  • High contrast dial
  • Contact point reverses automatically
  • Narrow body
  • Contact point included
  • Two dovetail clamps included
  • Case included
Catalog No. 50-005-13412 Supplier MSC Supplier No. 30752349
The Starrett Series 3909 Horizontal Dial Test Indicator is designed for precision measurements, featuring a robust gear-driven design and jeweled movement for accuracy. Its durable satin chrome finish ensures longevity, and the adjustable, replaceable contact point enhances versatility. With a high-contrast dial and automatic contact point reversal, this indicator provides reliable performance for various laboratory and industrial applications.
  • Precision gear-driven design
  • Jeweled movement
  • Satin chrome finish
  • Adjustable contact
  • Durable construction
  • Replaceable contact
  • High contrast dial
  • Contact point reverses automatically
  • Narrow body
  • Contact point included
  • Two dovetail clamps included
  • Case included