Learn More
MSC Starrett Individual Radius Gage, 0.020 in, Stainless Steel, Convex/Concave
Catalog No. 5000536901
Encompass
This individual radius gage is designed for precise measurement of 0.020 inch radii. Crafted from satin finish stainless steel, it is rust and stain resistant, ensuring durability and accuracy for various applications in industries such as aerospace, automotive, and metalworking.
  • Made of satin finish stainless steel.
  • Gage is clearly marked with its radius.
  • Features five different gaging surfaces for both convex and concave radii.
  • Precision finished radii with extra smooth, accurate edges.
Catalog No. 50-005-36901 Supplier MSC Supplier No. 58696279
This individual radius gage is designed for precise measurement of 0.020 inch radii. Crafted from satin finish stainless steel, it is rust and stain resistant, ensuring durability and accuracy for various applications in industries such as aerospace, automotive, and metalworking.
  • Made of satin finish stainless steel.
  • Gage is clearly marked with its radius.
  • Features five different gaging surfaces for both convex and concave radii.
  • Precision finished radii with extra smooth, accurate edges.