Learn More
MSC Starrett Inside Caliper, 12 Inch Leg, Spring Joint, Steel, Polished
Supplier:  MSC 86403581
Encompass
The product is a 12 Inch Leg, Spring Joint, Steel, Polished, Inside Caliper. It is designed for inside measurements and features flat legs and a solid nut type.
  • Spring joint for precise action.
  • Designed for inside measurements.
  • Flat leg design.
  • Solid nut for secure adjustments.
  • 12 inch leg length.
  • Screw adjustment method.
  • Constructed from steel for durability.
  • Polished finish.
  • Includes a screw.
  • Maximum measurement of 12 inches (300mm).
Catalog No. 50-005-31334