Learn More
MSC Starrett Inside Micrometer Set, Solid-Rod, 2-12 in Range, 0.001 in Graduations
Catalog No. 5000529988
Encompass
This solid-rod inside micrometer set is designed for precise measurements of inside diameters of cylinders, rings, and parallel surfaces. It offers lightness, ease of use, and a versatile measuring range, achieved by assembling rods and spacing collars to the micrometer head. Measuring rods feature a shoulder accurately set and locked in position for reliable readings.
  • Insulated rods marked with length
  • Rods are adjustable for length
  • Solid measuring rods
  • Hardened and ground anvils
  • Quick-reading figures
Catalog No. 50-005-29988 Supplier MSC Supplier No. 06358055
This solid-rod inside micrometer set is designed for precise measurements of inside diameters of cylinders, rings, and parallel surfaces. It offers lightness, ease of use, and a versatile measuring range, achieved by assembling rods and spacing collars to the micrometer head. Measuring rods feature a shoulder accurately set and locked in position for reliable readings.
  • Insulated rods marked with length
  • Rods are adjustable for length
  • Solid measuring rods
  • Hardened and ground anvils
  • Quick-reading figures