Learn More
MSC Starrett Interchangeable Anvil Micrometer, 0 mm to 100 mm Range, Ratchet Stop, 1 ea
Catalog No. 5000514128
Encompass
This mechanical interchangeable anvil micrometer features a 0 to 100 mm range, providing a versatile solution for applications requiring a single tool for a broad measurement capacity. Designed for precision in machine and automotive repair environments, it utilizes a series of interchangeable anvils to cover the full range in 25 mm increments.
  • Satin chrome finish resists rust and prevents glare for improved visibility.
  • Rigid special cast iron frame is ribbed for strength and perforated for a balanced, lightweight feel.
  • Advanced sleeve design with staggered lines and distinct figures ensures precise readability.
  • Combination ratchet and speeder allows for uniform measuring pressure and quick adjustments.
  • One-piece spindle construction ensures long-term accuracy and stability.
  • Includes a protective case and appropriate wrenches for adjustments.
Catalog No. 50-005-14128 Supplier MSC Supplier No. 58697889
This mechanical interchangeable anvil micrometer features a 0 to 100 mm range, providing a versatile solution for applications requiring a single tool for a broad measurement capacity. Designed for precision in machine and automotive repair environments, it utilizes a series of interchangeable anvils to cover the full range in 25 mm increments.
  • Satin chrome finish resists rust and prevents glare for improved visibility.
  • Rigid special cast iron frame is ribbed for strength and perforated for a balanced, lightweight feel.
  • Advanced sleeve design with staggered lines and distinct figures ensures precise readability.
  • Combination ratchet and speeder allows for uniform measuring pressure and quick adjustments.
  • One-piece spindle construction ensures long-term accuracy and stability.
  • Includes a protective case and appropriate wrenches for adjustments.