Learn More
MSC Starrett Last Word Dial Test Indicator, 0.7 mm Range, 0.002 mm Graduations
Catalog No. 5000535037
Encompass
This Last Word Dial Test Indicator is highly versatile, designed for precise measurements in all machining, layout, and inspection work. Its small size and various attachments ensure ease and accuracy. A shaded dial helps operators always know the correct reading, especially in applications like jig boring.
  • Small size and variety of attachments for all jobs.
  • Shaded dial for clear readings, even with a mirror.
  • Ideal for precise measurements in machining, layout, and inspection.
  • Easy reading dials with half yellow for clarity.
  • Hard chrome-plated ratchet contact point.
  • Smooth, jeweled lever action.
  • Positive reversing switch.
  • Indicator with body clamp only.
Catalog No. 50-005-35037 Supplier MSC Supplier No. 58706037
This Last Word Dial Test Indicator is highly versatile, designed for precise measurements in all machining, layout, and inspection work. Its small size and various attachments ensure ease and accuracy. A shaded dial helps operators always know the correct reading, especially in applications like jig boring.
  • Small size and variety of attachments for all jobs.
  • Shaded dial for clear readings, even with a mirror.
  • Ideal for precise measurements in machining, layout, and inspection.
  • Easy reading dials with half yellow for clarity.
  • Hard chrome-plated ratchet contact point.
  • Smooth, jeweled lever action.
  • Positive reversing switch.
  • Indicator with body clamp only.