Learn More
MSC Starrett Machinists Level, 18 in, Double Plumb Vial, Cross Test Vial
Catalog No. 5000531949
Encompass
The Starrett 98 Machinists Level, 18 inches long, features ground, graduated main vials, a cross test vial, and a double plumb vial for varied leveling tasks. Its base includes an involute groove for round work, while adjustable vials are housed in a satin finished brass tube with a friction-fit cover. Main vials provide graduations of approximately 80-90 seconds or .005 inches per foot (0.42mm per meter).
  • Ground and graduated main vials.
  • Involute groove in base for round work.
  • Includes cross test vial.
  • Adjustable vials with satin finished brass tube and friction-fit cover.
  • Main level graduations: 80-90 seconds or .005 inches per foot (0.42mm per meter).
  • Double plumb vial for increased versatility.
Catalog No. 50-005-31949 Supplier MSC Supplier No. 98144942
The Starrett 98 Machinists Level, 18 inches long, features ground, graduated main vials, a cross test vial, and a double plumb vial for varied leveling tasks. Its base includes an involute groove for round work, while adjustable vials are housed in a satin finished brass tube with a friction-fit cover. Main vials provide graduations of approximately 80-90 seconds or .005 inches per foot (0.42mm per meter).
  • Ground and graduated main vials.
  • Involute groove in base for round work.
  • Includes cross test vial.
  • Adjustable vials with satin finished brass tube and friction-fit cover.
  • Main level graduations: 80-90 seconds or .005 inches per foot (0.42mm per meter).
  • Double plumb vial for increased versatility.