Learn More
MSC Starrett Machinists Level, 18 in, Ground and Graduated Vial, Wood Case
Catalog No. 5000527211
Encompass
The Starrett 98Z-18 Machinists Level is a precision tool designed for accurate leveling in various applications. It features ground and graduated main vials, a double plumb vial, and a cross test vial for comprehensive measurements. The level, measuring 18 inches (approximately 450 mm) in length, comes with a protective wood case.
  • Ground and graduated main vials.
  • Base has an involute groove for stable seating on round work.
  • Cross test vial included.
  • Vials are adjustable for positive setting.
  • Vials are protected in a satin finished brass tube with a friction-fit cover.
  • Main level vials have graduations of approximately 80-90 seconds or .005 inch per foot (0.42 mm per meter).
  • Double plumb vial included.
  • Supplied with a wood case.
Catalog No. 50-005-27211 Supplier MSC Supplier No. 86404605
The Starrett 98Z-18 Machinists Level is a precision tool designed for accurate leveling in various applications. It features ground and graduated main vials, a double plumb vial, and a cross test vial for comprehensive measurements. The level, measuring 18 inches (approximately 450 mm) in length, comes with a protective wood case.
  • Ground and graduated main vials.
  • Base has an involute groove for stable seating on round work.
  • Cross test vial included.
  • Vials are adjustable for positive setting.
  • Vials are protected in a satin finished brass tube with a friction-fit cover.
  • Main level vials have graduations of approximately 80-90 seconds or .005 inch per foot (0.42 mm per meter).
  • Double plumb vial included.
  • Supplied with a wood case.