Learn More
MSC Starrett Machinists Level, 4 Inch, Cast Iron, Ground, Graduated Vials
Catalog No. 5000531167
Encompass
This precision machinists level is constructed from cast iron and features ground and graduated main vials for accurate measurements. It is designed for precise leveling in various applications.
  • Cast iron construction for durability.
  • Ground and graduated main vials for precise readings.
  • 4 inch length.
  • Base flatness of 1/64 "
  • Measures 7/8 inch wide by 1-3/16 inch high.
  • Graduation sensitivity of 0.0050 inch or 0.0500 inch per 10 inches.
Catalog No. 50-005-31167 Supplier MSC Supplier No. 90008228
This precision machinists level is constructed from cast iron and features ground and graduated main vials for accurate measurements. It is designed for precise leveling in various applications.
  • Cast iron construction for durability.
  • Ground and graduated main vials for precise readings.
  • 4 inch length.
  • Base flatness of 1/64 "
  • Measures 7/8 inch wide by 1-3/16 inch high.
  • Graduation sensitivity of 0.0050 inch or 0.0500 inch per 10 inches.