Learn More
MSC Starrett Mechanical Inside Micrometer Set, 2 to 8 in Range, 6 Extension Rods, Satin Chrome Finish
Catalog No. 5000527262
Encompass
This mechanical inside micrometer set is designed for precise internal measurements, offering a range of 2 to 8 inches. It includes six extension rods and features a satin chrome finish for durability. The set provides accurate readings with hardened and ground anvils, quick-reading figures, and insulated rods marked with length, all conveniently housed in a case.
  • Hardened and ground anvils
  • Quick-reading figures
  • Insulated rods marked with length
  • Measuring rods included
  • Comes with a protective case
  • 2 to 8 inch measurement range
  • 6 extension rods
  • Satin chrome finish
  • Solid rod type
  • Friction thimble type
  • Accuracy of 0.00010 inch
  • 1/2 inch travel
Catalog No. 50-005-27262 Supplier MSC Supplier No. 86405412
This mechanical inside micrometer set is designed for precise internal measurements, offering a range of 2 to 8 inches. It includes six extension rods and features a satin chrome finish for durability. The set provides accurate readings with hardened and ground anvils, quick-reading figures, and insulated rods marked with length, all conveniently housed in a case.
  • Hardened and ground anvils
  • Quick-reading figures
  • Insulated rods marked with length
  • Measuring rods included
  • Comes with a protective case
  • 2 to 8 inch measurement range
  • 6 extension rods
  • Satin chrome finish
  • Solid rod type
  • Friction thimble type
  • Accuracy of 0.00010 inch
  • 1/2 inch travel