Learn More
MSC Starrett Mechanical Inside Tubular Micrometer, 1.5-32 in Range, 0.001 in Graduation, 0.0001 in Accuracy
Catalog No. 5000531321
Encompass
The Starrett Mechanical Inside Tubular Micrometer is designed for precise internal measurements. It features a range of 1.5 to 32 inches, a graduation of 0.001 inch, and an accuracy of 0.0001 inch, making it suitable for demanding applications requiring high precision. This micrometer includes a friction thimble for consistent readings and a satin chrome finish for durability.
  • Hardened and ground anvils on rods are adjustable for length.
  • Friction thimble ensures consistent measurements.
  • Includes 10 extension rods for versatility.
  • Satin chrome finish provides enhanced durability.
Catalog No. 50-005-31321 Supplier MSC Supplier No. 86425063
The Starrett Mechanical Inside Tubular Micrometer is designed for precise internal measurements. It features a range of 1.5 to 32 inches, a graduation of 0.001 inch, and an accuracy of 0.0001 inch, making it suitable for demanding applications requiring high precision. This micrometer includes a friction thimble for consistent readings and a satin chrome finish for durability.
  • Hardened and ground anvils on rods are adjustable for length.
  • Friction thimble ensures consistent measurements.
  • Includes 10 extension rods for versatility.
  • Satin chrome finish provides enhanced durability.