Learn More
MSC Starrett Mechanical Interchangeable Anvil Micrometer, 12 to 18 in Range, 5 Anvils
Catalog No. 5000511808
Encompass
The Starrett Mechanical Interchangeable Anvil Micrometer is engineered for accurate and reliable measurements in demanding environments. This precision instrument offers a measuring range of 12 to 18 inches and is equipped with 5 interchangeable anvils for enhanced versatility. Its mechanical operation and friction thimble provide consistent performance for expert users.
  • Mechanical operation for consistent performance.
  • Measurement range of 12 to 18 inches.
  • Includes 5 anvils for versatile applications.
  • Features a friction thimble for precise control.
  • Accuracy of 0.0003 inch for reliable readings.
  • Equipped with a ring lock nut for secure measurements.
  • Constructed with steel anvils and spindle for durability.
  • Comes with a wrench for adjustments and maintenance.
Catalog No. 50-005-11808 Supplier MSC Supplier No. 86424538
The Starrett Mechanical Interchangeable Anvil Micrometer is engineered for accurate and reliable measurements in demanding environments. This precision instrument offers a measuring range of 12 to 18 inches and is equipped with 5 interchangeable anvils for enhanced versatility. Its mechanical operation and friction thimble provide consistent performance for expert users.
  • Mechanical operation for consistent performance.
  • Measurement range of 12 to 18 inches.
  • Includes 5 anvils for versatile applications.
  • Features a friction thimble for precise control.
  • Accuracy of 0.0003 inch for reliable readings.
  • Equipped with a ring lock nut for secure measurements.
  • Constructed with steel anvils and spindle for durability.
  • Comes with a wrench for adjustments and maintenance.