Learn More
MSC Starrett Mechanical Outside Micrometer Set, 0-4 in Range, 0.0001 in Graduations, 4 Piece
Catalog No. 5000527078
Encompass
This mechanical outside micrometer set includes four micrometers, providing a 0-4 in measurement range with 0.0001 in graduations. It features a heat-insulator on the frame to minimize temperature-related expansion or contraction. The spindle and anvil have flat, carbide-tipped measuring faces for wear resistance, and a spindle lock ensures secure measurement locking. A combination ratchet and speeder allows for consistent pressure and faster adjustments.
  • Accurate, rugged, and easy to use.
  • Ratchet stop, lock nut, and standards included.
  • Micro-lapped measuring faces.
  • No-glare satin chrome finish on thimble and sleeve minimizes glare and resists rust.
  • Protective case keeps micrometers and standards organized and accessible.
  • Balanced frame and thimble design for easy handling.
  • Ring-type knurled lock nut for quick and secure locking.
Catalog No. 50-005-27078 Supplier MSC Supplier No. 48864433
This mechanical outside micrometer set includes four micrometers, providing a 0-4 in measurement range with 0.0001 in graduations. It features a heat-insulator on the frame to minimize temperature-related expansion or contraction. The spindle and anvil have flat, carbide-tipped measuring faces for wear resistance, and a spindle lock ensures secure measurement locking. A combination ratchet and speeder allows for consistent pressure and faster adjustments.
  • Accurate, rugged, and easy to use.
  • Ratchet stop, lock nut, and standards included.
  • Micro-lapped measuring faces.
  • No-glare satin chrome finish on thimble and sleeve minimizes glare and resists rust.
  • Protective case keeps micrometers and standards organized and accessible.
  • Balanced frame and thimble design for easy handling.
  • Ring-type knurled lock nut for quick and secure locking.