Learn More
MSC Starrett Mechanical Outside Micrometer, 0 to 13 mm Range, 0.01 mm Graduation, 1 ea.
Catalog No. 5000510652
Encompass
The mechanical outside micrometer is a precision measuring tool designed for accurate dimensional inspection. Featuring a 0 mm to 13 mm measuring range and 0.01 mm graduation, this instrument provides reliable readings for metalworking and engineering applications.
  • Graduation of 0.01 mm for precise measurements
  • Measuring range of 0 mm to 13 mm
  • Spindle and anvil size of 0.235 in.
  • Choice of smooth friction thimble for uniform pressure
  • Choice of combination ratchet and speeder for uniform pressure and quicker adjustment
Catalog No. 50-005-10652 Supplier MSC Supplier No. 86409273
The mechanical outside micrometer is a precision measuring tool designed for accurate dimensional inspection. Featuring a 0 mm to 13 mm measuring range and 0.01 mm graduation, this instrument provides reliable readings for metalworking and engineering applications.
  • Graduation of 0.01 mm for precise measurements
  • Measuring range of 0 mm to 13 mm
  • Spindle and anvil size of 0.235 in.
  • Choice of smooth friction thimble for uniform pressure
  • Choice of combination ratchet and speeder for uniform pressure and quicker adjustment