Learn More
MSC Starrett Micrometer Depth Gage, 0-150mm, Non-Rotating Blade, 63.5mm Base
Catalog No. 5000537084
Encompass
This depth micrometer features a non-rotating blade, providing precise depth measurements in challenging applications. With a 0-150mm range and 0.01mm graduation, it is suitable for measuring narrow shoulders, slots, and recesses. The blade, measuring 1.2 mm thick by 3.2 mm wide (0.045 x 1/8 in), moves perpendicularly, ensuring accurate readings without rolling off.
  • Non-rotating blade moves perpendicularly to prevent rolling off narrow shoulders
  • Ideal for measuring depths of narrow shoulders, slots, and recesses as narrow as 1.2 mm (0.045 in)
  • Blade can be positioned at any angle relative to the base
  • Includes a 63.5 mm (2-1/2 in) base
  • Features a combination ratchet and speeder for uniform pressure and quicker adjustment
  • Equipped with a ring-type knurled lock nut for quick and secure locking
  • Comes with 6 measuring rods
Catalog No. 50-005-37084 Supplier MSC Supplier No. 58702523
This depth micrometer features a non-rotating blade, providing precise depth measurements in challenging applications. With a 0-150mm range and 0.01mm graduation, it is suitable for measuring narrow shoulders, slots, and recesses. The blade, measuring 1.2 mm thick by 3.2 mm wide (0.045 x 1/8 in), moves perpendicularly, ensuring accurate readings without rolling off.
  • Non-rotating blade moves perpendicularly to prevent rolling off narrow shoulders
  • Ideal for measuring depths of narrow shoulders, slots, and recesses as narrow as 1.2 mm (0.045 in)
  • Blade can be positioned at any angle relative to the base
  • Includes a 63.5 mm (2-1/2 in) base
  • Features a combination ratchet and speeder for uniform pressure and quicker adjustment
  • Equipped with a ring-type knurled lock nut for quick and secure locking
  • Comes with 6 measuring rods