Learn More
MSC Starrett Micrometer Standard Set, 12 Pieces, 12-23 in Range, Spherical End
Catalog No. 5000534893
Encompass
This micrometer standard set includes 12 pieces designed for checking and setting micrometers with a 12-23 inch range. These standards are crafted from special tool steel with hardened ends accurately lapped to a spherical radius. They are suitable for various applications including comparing gages, checking precision measuring tools, and measuring parallel surfaces.
  • Designed for checking and setting micrometers
  • Useful on machine tools for comparing gages
  • Ideal for checking precision measuring tools
  • Suitable for measuring parallel surfaces
  • Constructed from special tool steel in rod form
  • Ends are hardened and accurately lapped to a spherical radius
  • Set provides 12 individual standards
  • Covers a length range from 12 to 23 inches
Catalog No. 50-005-34893 Supplier MSC Supplier No. 58701996
This micrometer standard set includes 12 pieces designed for checking and setting micrometers with a 12-23 inch range. These standards are crafted from special tool steel with hardened ends accurately lapped to a spherical radius. They are suitable for various applications including comparing gages, checking precision measuring tools, and measuring parallel surfaces.
  • Designed for checking and setting micrometers
  • Useful on machine tools for comparing gages
  • Ideal for checking precision measuring tools
  • Suitable for measuring parallel surfaces
  • Constructed from special tool steel in rod form
  • Ends are hardened and accurately lapped to a spherical radius
  • Set provides 12 individual standards
  • Covers a length range from 12 to 23 inches