Learn More
MSC Starrett Outside Micrometer, 4-5 inch Range, 0.0001 inch Graduation, Carbide Anvil
Catalog No. 5000510577
Encompass
These outside micrometers are widely used globally for their accuracy, ruggedness, and ease of use. This model features a 4-5 inch range, 0.0001 inch graduations, a flat carbide anvil, a knurled ring type locknut, and a ratchet thimble. It comes with a black wrinkle frame finish and a storage case.
  • Popular precision micrometers used worldwide.
  • Accurate, rugged, and easy to use.
  • Satin chrome finish is glare-free and resists rust.
  • Advanced sleeve design with staggered lines and distinct figures for precise and easy readability.
  • Balanced frame and thimble design for easy handling.
  • Gracefully designed tapered frame for use in narrow slots and tight places.
  • Quick and easy adjustment.
  • Micro-lapped measuring faces.
  • Extremely hard and stable one-piece spindle.
  • Made in the USA with US and global content.
Catalog No. 50-005-10577 Supplier MSC Supplier No. 85549236
These outside micrometers are widely used globally for their accuracy, ruggedness, and ease of use. This model features a 4-5 inch range, 0.0001 inch graduations, a flat carbide anvil, a knurled ring type locknut, and a ratchet thimble. It comes with a black wrinkle frame finish and a storage case.
  • Popular precision micrometers used worldwide.
  • Accurate, rugged, and easy to use.
  • Satin chrome finish is glare-free and resists rust.
  • Advanced sleeve design with staggered lines and distinct figures for precise and easy readability.
  • Balanced frame and thimble design for easy handling.
  • Gracefully designed tapered frame for use in narrow slots and tight places.
  • Quick and easy adjustment.
  • Micro-lapped measuring faces.
  • Extremely hard and stable one-piece spindle.
  • Made in the USA with US and global content.