Learn More
MSC Starrett 232 Series Mechanical Outside Micrometer, 0-1/2 in. Range, 0.0001 in. Resolution, 1 ea.
Catalog No. 5000515302
Encompass
The 232 Series mechanical outside micrometer is a precision instrument designed for accurate measurements in machining and inspection applications. Featuring a 0 to 1/2 in. measuring range, this tool utilizes a rigid, drop-forged steel frame and carbide-tipped measuring faces to ensure durability and stability during use.
  • Advanced sleeve design with staggered lines and distinct figures for precise, easy readability
  • Balanced frame and thimble design for easy handling
  • Tapered frame for use in narrow slots and tight places
  • Rust and glare-resistant satin chrome finish
  • Combination ratchet and speeder for uniform pressure and quick adjustment
  • Ring-type knurled lock nut for secure locking
  • Convenient decimal equivalents on the frame
Catalog No. 50-005-15302 Supplier MSC Supplier No. 86409281
The 232 Series mechanical outside micrometer is a precision instrument designed for accurate measurements in machining and inspection applications. Featuring a 0 to 1/2 in. measuring range, this tool utilizes a rigid, drop-forged steel frame and carbide-tipped measuring faces to ensure durability and stability during use.
  • Advanced sleeve design with staggered lines and distinct figures for precise, easy readability
  • Balanced frame and thimble design for easy handling
  • Tapered frame for use in narrow slots and tight places
  • Rust and glare-resistant satin chrome finish
  • Combination ratchet and speeder for uniform pressure and quick adjustment
  • Ring-type knurled lock nut for secure locking
  • Convenient decimal equivalents on the frame