Learn More
MSC Starrett Parallel, Tool Steel, Four Face, 0.0001 in Parallelism
Catalog No. 5000535233
Encompass
This parallel is made from a special grade of tool steel and is ground to close limits, ensuring exceptional accuracy with 0.0001 inch parallelism. Designed as a four-face parallel, it measures 6 inches long, 1 inch high, and 1/2 inch thick, and is sold as a matched pair for comprehensive precision applications.
  • Made from a special grade of tool steel.
  • Ground to close limits.
  • Four face design.
  • Features 0.0001 inch parallelism.
  • 6 inches (152.40 mm) long.
  • 1 inch (25.40 mm) high.
  • 1/2 inch (12.70 mm) thick.
  • Four precision-ground sides.
  • Sold as a matched pair.
Catalog No. 50-005-35233 Supplier MSC Supplier No. 58700121
This parallel is made from a special grade of tool steel and is ground to close limits, ensuring exceptional accuracy with 0.0001 inch parallelism. Designed as a four-face parallel, it measures 6 inches long, 1 inch high, and 1/2 inch thick, and is sold as a matched pair for comprehensive precision applications.
  • Made from a special grade of tool steel.
  • Ground to close limits.
  • Four face design.
  • Features 0.0001 inch parallelism.
  • 6 inches (152.40 mm) long.
  • 1 inch (25.40 mm) high.
  • 1/2 inch (12.70 mm) thick.
  • Four precision-ground sides.
  • Sold as a matched pair.