Learn More
MSC Starrett Trammel Points, Forged Steel, 5 in. Length, 2/Pk.
Catalog No. 5000510616
Encompass
These 5 in. (125 mm) trammel points are precision engineered from forged steel to provide durable and accurate marking for layout applications. Designed for compatibility with the 50B trammel bar, they offer a reliable solution for extending the capacity of layout tools in machine shop and metalworking environments.
  • Constructed from hardened forged steel
  • Features a durable wrinkle finish
  • Includes two points for use with 50B models
  • Designed for 5 in. leg length requirements
Catalog No. 50-005-10616 Supplier MSC Supplier No. 86402971
These 5 in. (125 mm) trammel points are precision engineered from forged steel to provide durable and accurate marking for layout applications. Designed for compatibility with the 50B trammel bar, they offer a reliable solution for extending the capacity of layout tools in machine shop and metalworking environments.
  • Constructed from hardened forged steel
  • Features a durable wrinkle finish
  • Includes two points for use with 50B models
  • Designed for 5 in. leg length requirements