Learn More
MSC Starrett Precision Steel Pin Gage Set, .251-.500 in, 0.0002 in Accuracy
Catalog No. 5000534373
Encompass
The S4000 Precision Steel Pin Gage Set is designed for accurately determining small hole sizes, gaging slots, and finding hole distances. This set covers a range from .251 to .500 inches and includes 250 gages made from hardened tool steel, manufactured to a 0.0002 inch tolerance.
  • Color coded, fully adjustable Go/No-Go gage handle is included.
  • Supplied in rugged, high-impact protective cases with marked spaces for each gage.
  • An inspection certificate is provided with every set.
  • Pin gages are manufactured to a 0.0002 inch tolerance.
  • Plus and minus tolerance sets are available.
  • Offered in 0.001 inch increments.
  • Each pin is centerless lapped and clearly etched with the stated size.
  • Gages are 2 inches long and hardened to RC 60/66.
  • All sharp corners are broken.
Catalog No. 50-005-34373 Supplier MSC Supplier No. 88175385
The S4000 Precision Steel Pin Gage Set is designed for accurately determining small hole sizes, gaging slots, and finding hole distances. This set covers a range from .251 to .500 inches and includes 250 gages made from hardened tool steel, manufactured to a 0.0002 inch tolerance.
  • Color coded, fully adjustable Go/No-Go gage handle is included.
  • Supplied in rugged, high-impact protective cases with marked spaces for each gage.
  • An inspection certificate is provided with every set.
  • Pin gages are manufactured to a 0.0002 inch tolerance.
  • Plus and minus tolerance sets are available.
  • Offered in 0.001 inch increments.
  • Each pin is centerless lapped and clearly etched with the stated size.
  • Gages are 2 inches long and hardened to RC 60/66.
  • All sharp corners are broken.