Learn More
MSC Starrett Radius Gage, 1/16 Inch Radius, Satin Chrome Finish, Stainless Steel
Catalog No. 5000530220
Encompass
This radius gage, manufactured by Starrett, is designed for precise measurement of both convex and concave radii. Constructed from durable stainless steel with a satin chrome finish, it ensures accuracy and longevity in various applications. Each leaf is configured to measure both convex and concave radii, providing versatility for detailed work.
  • Measures 1/16 inch radius.
  • Features a maximum radius of 0.06 "
  • Configured for both convex and concave radii measurements per leaf.
  • Constructed from stainless steel for durability.
  • Finished with satin chrome for enhanced resilience and visibility.
Catalog No. 50-005-30220 Supplier MSC Supplier No. 86406287
This radius gage, manufactured by Starrett, is designed for precise measurement of both convex and concave radii. Constructed from durable stainless steel with a satin chrome finish, it ensures accuracy and longevity in various applications. Each leaf is configured to measure both convex and concave radii, providing versatility for detailed work.
  • Measures 1/16 inch radius.
  • Features a maximum radius of 0.06 "
  • Configured for both convex and concave radii measurements per leaf.
  • Constructed from stainless steel for durability.
  • Finished with satin chrome for enhanced resilience and visibility.